Cole-Parmer® PCR Tubes
Avoid solvent evaporation with efficient heat transfer PCR tubes
  • PCR products are manufactured from prime virgin polypropylene
  • Pair directly with your 50ml centrifuge tubes
  • 0.2ml thin wall PCR tubes, strip of 8 tubes, flat caps
  • Ultra-thin wall design for efficient heat transfer
  • Compatible with standard 96-well heat blocks
  • High light transmittance
  • Sterilized
  • Packaged in easy opening plastic bag and white box
  • Inner packaging is individually labelled for lot traceability
5,900.00 - ₹80,400.00INR / Case of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
125 tubes/bag; 10 bags/case. Total: 1250 tubes
₹80,400.00INR / Case of 1
COMPARE
1000 tubes/bag; 10 bags/case. Total: 10000 tubes
₹59,900.00INR / Case of 1
COMPARE
Flat Cap Tubes, Clear, 0.2 mL; 500/Pk, 6 Pks/Cs
₹8,000.00INR / Case of 3000
COMPARE
8-STRIP Tubes with joint Flat Caps, 0.2 mL; 12/Pk, 10 Pk/Cs
₹5,900.00INR / Case of 120
COMPARE
8-STRIP Tubes with Flat Caps, 0.2 mL, 12/Pk, 10 Pk/Cs
₹6,700.00INR / Case of 120
COMPARE
8-STRIP Tubes with Domed Caps, 0.2 mL, 12/Pk, 10 Pk/Cs
₹7,700.00INR / Case of 120
COMPARE
QPCR 8-STRIP Tubes with joint Flat Caps, 0.1 mL, White; 12/Pk, 10 Pk/Cs
₹9,100.00INR / Case of 120
COMPARE
QPCR 8-STRIP Tubes with Optical Caps, 0.1 mL, White; 12/Pk, 10 Pk/Cs
₹9,900.00INR / Case of 120
COMPARE
QPCR 8-STRIP Tubes with Optical Caps, 0.2 mL; 12/Pk, 10 Pk/Cs
₹9,100.00INR / Case of 120
COMPARE
QPCR 8-STRIP Tubes with Optical Caps, 0.2 mL, White; 12/Pk, 10 Pk/Cs
₹9,900.00INR / Case of 120
Showing 1- 10 of 10