Cole-Parmer 0.2ml PCR Tubes with Flat Caps 8 Strips Sterile(125/Bag,10 Bags/Case)
Representative image only.
Cole-Parmer 0.2ml PCR Tubes with Flat Caps 8 Strips Sterile(125/Bag,10 Bags/Case)
Cole-Parmer | Item: 1597276
Avoid solvent evaporation with efficient heat transfer PCR tubes
₹80,400.00INR / Case of 1
₹80,400.00INR / Case of 1
Product Description
Key Features
  • PCR products are manufactured from prime virgin polypropylene
  • Pair directly with your 50ml centrifuge tubes
  • 0.2ml thin wall PCR tubes, strip of 8 tubes, flat caps
  • Ultra-thin wall design for efficient heat transfer
  • Compatible with standard 96-well heat blocks
  • High light transmittance
  • Sterilized
  • Packaged in easy opening plastic bag and white box
  • Inner packaging is individually labelled for lot traceability
Specifications
  • Capacity0.2ml, 8 strips
  • Description125 tubes/bag; 10 bags/case. Total: 1250 tubes