Cole-Parmer® PCR Plates and Membranes
Designed to amplify nucleic acids during the PCR process
  • Precision molded from 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable.
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
Cole-Parmer PCR products are specifically designed for amplifying nucleic acids during the Polymerase Chain Reaction (PCR) process. Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.
7,700.00 - ₹35,600.00INR / Pkg of 50
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
96 Well PCR Plate, 0.2 mL, No Skirt, Standard; 50/Pk
₹10,500.00INR / Pkg of 50
COMPARE
96 Well PCR Plate, 0.2 mL, Sub-Semi Skirted, Standard; 50/Pk
₹16,300.00INR / Pkg of 50
COMPARE
96 Well QPCR Plate, 0.1 mL, Sub-Semi Skirted, Low Profile; 50/Pk
₹16,300.00INR / Pkg of 50
COMPARE
96 Well QPCR Plate, 0.1 mL, Semi Skirted, Low Profile; 50/Pk
₹16,300.00INR / Pkg of 50
COMPARE
96 Well QPCR Plate, 0.1 mL, Semi Skirted, White, Low Profile; 50/Pk
₹18,300.00INR / Pkg of 50
COMPARE
Sealing Membrane for PCR 96 Well Plate; 100/Pk
₹7,700.00INR / Pkg of 100
COMPARE
Presure sensitive membrane for QPCR 96 well Optical Sealing; 100/Pk
₹35,600.00INR / Pkg of 100
COMPARE
Aluminium Membrane; 100/Pk
₹7,700.00INR / Pkg of 100
Showing 1- 8 of 8