Cole-Parmer Sealing Membrane for PCR 96 Well Plate; 100/PK
Representative image only.
Cole-Parmer Sealing Membrane for PCR 96 Well Plate; 100/PK
Cole-Parmer | Item: 1597819
Designed to amplify nucleic acids during the PCR process
₹7,700.00INR / Pkg of 100
₹7,700.00INR / Pkg of 100
Product Description
Key Features
  • Precision molded from 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable.
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
  • 1 year warranty
More About this Item
Cole-Parmer PCR products are specifically designed for amplifying nucleic acids during the Polymerase Chain Reaction (PCR) process. Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.
1 Year Warranty
Specifications
  • SterileNon-Sterile
  • MaterialPolypropylene
  • DescriptionSealing Membrane for PCR 96 Well Plate; 100/Pk