Cole-Parmer OVF-800 Series Mechanical Convection Ovens
Optimized airflow provides even heat transfer for high temperature uniformity
  • Three-point temperature calibration ensures excellent temperature accuracy over a wide temperature range
  • Intuitive 3.5” touch screen controller
  • Stainless steel interior resists corrosion
  • Wide-latch handle makes it easy to open and close door even when hands are full
  • Stay-cool exterior for user safety
Use these fast-drying, ultra-magnetic ovens for thermal curing and denaturation of samples, preheating of building materials and electronic components, and experimental heat resistance. A microprocessor PID controller provides accurate and reliable temperature control up to 482°F (250°C). An internal fan quickly, evenly, and precisely distributes the heat to heat the chamber.
View current and set temperatures, timer setting, operating status, alarm conditions, and heat status on a 3.5” LCD. Touch screen makes it simple to set the target temperature and time. Use the timer to schedule start and end times as well as to maintain a temperature for a set time. Controller automatically records 12 most recent events, such as start/stop of operation and any error occurrence.
Mineral wool resin insulation and high-temperature silicone rubber gasket on the door ensure excellent insulation and minimal heat loss. Safety features include overtemperature protection, fan and heater shut-off when the door opens, and stay-cool exterior. The auto-run function saves the previous temperature settings in case of a power outage. Side ventilation port (33 mm dia) lets you control the amount of air to be exhausted. Password protection prevents any accidental setting changes.
476,000.00 - ₹686,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Power (VAC) Temperature Uniformity
Availability Pricing
COMPARE
26 230 ±1.4°C (±0.36°F)
₹476,000.00INR / Each
COMPARE
26 120 ±1.4°C (±0.36°F)
₹476,000.00INR / Each
COMPARE
61 230 ±1.9°C (±2.52°F)
₹491,000.00INR / Each
COMPARE
61 120 ±1.9°C (±2.52°F)
₹491,000.00INR / Each
COMPARE
112 230 ±1.7°C (±2.16°F)
₹609,000.00INR / Each
COMPARE
112 120 ±1.7°C (±2.16°F)
₹609,000.00INR / Each
COMPARE
147 230 ±1.5°C (±1.8°F)
₹686,000.00INR / Each
COMPARE
147 120 ±1.5°C (±1.8°F)
₹686,000.00INR / Each
Showing 1- 8 of 8