Cole-Parmer® Muffle Furnaces with drop down & side swing door and brick insulation
Designed for occasional intermittent work
  • Available in 5 capacities
  • Drop down door can be used as a work platform and side swing door is for user safety
  • Double Low thermal mass insulation for low external housing temperature
  • Built with low density ceramic bricks and ceramic fiber for faster heating rate
Design is compact and lightweight, and consists of 2 or 3 heating plates with Kanthal® AF. Metal case with chrome-phosphatizing base protection and external finish with heat-resistant metal paint. Easy loading drop down door and side swing door close hermetically, brick on brick. Furnace has a ceramic chimney.
These furnaces have lower front or lateral control panel, solid state relay. Each model features PAD Digital control with microprocessor, PID parameters, non-volatile memory and alarm. Different control options with multiple programming and has data logger and programmer communication by Ethernet/RS-232. These furnaces feature safety shut-off for thermocouple failure. Different range of accessories available as per the application need.
377,000.00 - ₹916,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Muffle Furnace, 1.6 L, 1200 C, Drop down and side swing door; 1/Ea
₹377,000.00INR / Each
COMPARE
Muffle Furnace, 7.6 L, 1200 C, Drop down and side swing door; 1/Ea
₹470,000.00INR / Each
COMPARE
Muffle Furnace, 18 L, 1200 C, Drop down and side swing door; 1/Ea
₹618,000.00INR / Each
COMPARE
Muffle Furnace, 24 L, 1200 C, Drop down and side swing door; 1/Ea
₹660,000.00INR / Each
COMPARE
Muffle Furnace, 28 L, 1200 C, Drop down and side swing door; 1/Ea
₹916,000.00INR / Each