Cole-Parmer® MicroFiber Wiper
Ultraclean and highly sorbent wiper ideal for critical environments
  • Ultrasonic sealed edge ensures ultra-low fiber release
  • Laundered and packed in cleanroom environment
  • Double bagged and vacuum packed for cleanroom passage
  • Twisted fiber prevents fiber shedding during rigorous wiping
  • Tightly woven, tortuous surfaces absorb high levels of water and oil
Cole-Parmer® microfiber wipers are 100% continuous polyester and polyamide micro denier filament woven wiper with twisted fiber — ideal to be used for applications requiring critical control of contaminants and around rough & abrasive areas without shedding fibers. Micro-denier substrates provide deep cleaning due to their advanced structure where particles are trapped inside and between the fibers.
Applications include:
• Designed for use in Class 10 (ISO Class 4) and higher cleanroom environments
• Cleaning or disinfecting workstation surfaces & processing equipment
• Cleaning optical machinery, flat panel displays and tools
• Applying or removing cleaning and disinfecting solutions
• Cleaning of medical device products
• Cleaning delicate or scratch sensitive surfaces
• Absorbing spills of most common disinfectants
21,800.00 - ₹48,700.00INR / Case of 1400
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Cole-Parmer Microfiber Wiper, 9" x 9"; 140 Pcs/Bag, 10 Bags/Case
₹30,100.00INR / Case of 1400
COMPARE
Cole-Parmer Microfiber Wiper, 12" X 12"; 100 Pcs/Bag, 10 Bags/Case
₹21,800.00INR / Case of 1000
COMPARE
Cole-Parmer Microfiber wiper,9" x 9",Gamma sterilized; 1400/CsMicrofiber Wiper, 9" x 9", Gamma sterilized; 140 Pcs/Bag, 10 Bags/Case
₹48,700.00INR / Case of 1400
COMPARE
Cole-Parmer Microfiber Wiper, 12" X 12", Gamma sterilized; 100 Pcs/Bag, 10 Bags/Case
₹38,700.00INR / Case of 1000