Cole-Parmer Microfiber Wiper, 12" X 12", Gamma sterilized; 100 Pcs/Bag, 10 Bags/Case
Representative image only.
Cole-Parmer Microfiber Wiper, 12" X 12", Gamma sterilized; 100 Pcs/Bag, 10 Bags/Case
Cole-Parmer | Item: 1597666
Ultraclean and highly sorbent wiper ideal for critical environments
₹38,700.00INR / Case of 1000
₹38,700.00INR / Case of 1000
Buy It Together
Current Item
Cole-Parmer Microfiber Wiper, 12" X 12", Gamma sterilized; 100 Pcs/Bag, 10 Bags/Case
Cole-Parmer Microfiber Wiper, 12" X 12", Gamma sterilized; 100 Pcs/Bag, 10 Bags/Case
1597666
₹38,700.00INR / Case of 1000
+
Diversey Oxivir® Tb Disinfectant Solution, 55 Gallon Drum
7890086
₹622,000.00INR / Each
=
Total Price 660700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ultrasonic sealed edge ensures ultra-low fiber release
  • Laundered and packed in cleanroom environment
  • Double bagged and vacuum packed for cleanroom passage
  • Twisted fiber prevents fiber shedding during rigorous wiping
  • Tightly woven, tortuous surfaces absorb high levels of water and oil
More About this Item
Cole-Parmer® microfiber wipers are 100% continuous polyester and polyamide micro denier filament woven wiper with twisted fiber — ideal to be used for applications requiring critical control of contaminants and around rough & abrasive areas without shedding fibers. Micro-denier substrates provide deep cleaning due to their advanced structure where particles are trapped inside and between the fibers.
Applications include:
• Designed for use in Class 10 (ISO Class 4) and higher cleanroom environments
• Cleaning or disinfecting workstation surfaces & processing equipment
• Cleaning optical machinery, flat panel displays and tools
• Applying or removing cleaning and disinfecting solutions
• Cleaning of medical device products
• Cleaning delicate or scratch sensitive surfaces
• Absorbing spills of most common disinfectants
Specifications
  • DescriptionCole-Parmer Microfiber Wiper, 12" X 12", Gamma sterilized; 100 Pcs/Bag, 10 Bags/Case