Cole-Parmer® Essentials High Alumina Cylindrical Crucible
Withstand temperatures up to 3182°F
  • Autoclavable
Unglazed cylindrical crucibles are 99.7% pure aluminum oxide. Withstand temperatures up to 3182°F (1750°C).
4,800.00 - ₹9,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
10 28
₹4,800.00INR / Each
COMPARE
20 32
₹6,600.00INR / Each
COMPARE
30 35
₹7,800.00INR / Each
COMPARE
50 41
₹9,300.00INR / Each