Cole-Parmer® Full Height Sieves, Stainless Steel, 3" dia
Made of stainless steel (SS)
  • Individually serial numbered
  • 1" stacking height
  • Compliant with ASTM E11 requirements for compliance sieves
These test sieves feature a stainless steel woven-wire cloth available wtih various openings and a full height stainless steel frame. Ideal for use with Cole-Parmer sieve shaker (59986-01), sold separately.
Optional accessories include a stainless steel cover (59988-03) and full-height receiving pan (59988-07), sold separately.
15,500.00 - ₹25,200.00INR / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (InchShort) Sieve Opening (mm) U.S. Standard Screen Size (No.)
Availability Pricing
COMPARE
1 4 5
₹15,500.00INR / Each
COMPARE
1 3.35 6
₹15,500.00INR / Each
COMPARE
1 2.83 7
₹15,500.00INR / Each
COMPARE
1 2.36 8
₹15,500.00INR / Each
COMPARE
1 2 10
₹15,500.00INR / Each
COMPARE
1 1.68 12
₹15,500.00INR / Each
COMPARE
1 1.41 14
₹15,500.00INR / Each
COMPARE
1 1.19 16
₹15,500.00INR / Each
COMPARE
1 1 18
₹15,500.00INR / Each
COMPARE
1 0.841 20
₹15,500.00INR / Each
COMPARE
1 0.707 25
₹15,500.00INR / Each
COMPARE
1 0.595 30
₹15,500.00INR / Each
COMPARE
1 0.5 35
₹15,500.00INR / Each
COMPARE
1 0.425 40
₹15,500.00INR / Each
COMPARE
1 0.355 45
₹15,500.00INR / Each
COMPARE
1 0.297 50
₹15,500.00INR / Each
COMPARE
1 0.25 60
₹15,500.00INR / Each
COMPARE
1 0.212 70
₹15,500.00INR / Each
COMPARE
1 0.177 80
₹15,500.00INR / Each
COMPARE
1 0.149 100
₹15,500.00INR / Each
Showing 1 - 20 of 28