Cole-Parmer® Fluorescent Yellow/Green Dye Tracers
Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
8,600.00 - ₹29,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Fluorescent Flt Yellowith Green Dye Tablets, 200 Tablets
₹12,800.00INR / Each
COMPARE
Fluorescent Flt Yellowith Green Dye Concentrate, Gallon
₹27,800.00INR / Gallon of 1
COMPARE
Fluorescent Flt Yellowith Green Dye Concentrate, Pint
₹8,600.00INR / Pint of 1
COMPARE
Fluorescent Flt Yellow with Green Dye Powder, 1 Lb
₹29,700.00INR / Each