Cole-Parmer Fluorescent Flt Yellowith Green Dye Concentrate, Gallon
Representative image only.
Cole-Parmer Fluorescent Flt Yellowith Green Dye Concentrate, Gallon
Cole-Parmer | Item: 0029807
Determine water flow in closed circuits or open channels
₹27,800.00INR / Gallon of 1
₹27,800.00INR / Gallon of 1
Product Description
Key Features
  • Biodegradable and nontoxic
More About this Item
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
NSF international certified
Specifications
  • Reagent PlatformLiquid concentrate
  • Gallons Of Water For 1 ppm (Gal)100000
  • DescriptionFluorescent Flt Yellowith Green Dye Concentrate, Gallon