Cole-Parmer Fluorescent Flt Yellow/Green Dye Tablets, 200 Tablets
Representative image only.
Cole-Parmer Fluorescent Flt Yellow/Green Dye Tablets, 200 Tablets
Cole-Parmer | Item: 0029517
Determine water flow in closed circuits or open channels
₹12,800.00INR / Each
₹12,800.00INR / Each
Product Description
Key Features
  • Biodegradable and nontoxic
More About this Item
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.
NSF international certified
Specifications
  • Reagent PlatformTablet
  • Gallons Of Water For 1 ppm (Gal)60
  • Tests Per Kit200
  • DescriptionFluorescent Flt Yellowith Green Dye Tablets, 200 Tablets