Cole-Parmer® Fixed-Position Utility Clamps
Clamps include a built-in holder that grips rods up to ¾” (19mm)
  • An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
Used to hold apparatus at a fixed 90° angle near the lab frame where no adjustment is required after set-up. Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Utilizing thumbscrews, both prongs are easily adjusted over a wider range. Offered in either stainless steel or nickel plated zinc. Supplied with non-slip vinyl sleeves or for temperatures above 212°F (100°C), fiberglass covers.
7,000.00 - ₹42,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Accepts Rod Diameter (in) Clamp Type Length (in)
Availability Pricing
COMPARE
0.75 2-Prong Single Adjust Nickel-Plated Zinc 5.24
₹7,000.00INR / Each
COMPARE
0.75 3-Prong Dual Adjust Nickel-Plated Zinc 5.75
₹8,400.00INR / Each
COMPARE
0.75 3-Prong Dual Adjust Stainless Steel 5.75
₹32,300.00INR / Each
COMPARE
0.75 2-Prong Single Adjust Stainless Steel 5.24
₹42,400.00INR / Each