Cole-Parmer Buret Clamp, Three Prongs, Dual Adjustment, Medium
Representative image only.
Cole-Parmer Buret Clamp, Three Prongs, Dual Adjustment, Medium
Cole-Parmer | Item: 0802202
Clamps include a built-in holder that grips rods up to ¾” (19mm)
₹8,400.00INR / Each
₹8,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Buret Clamp, Three Prongs, Dual Adjustment, Medium
Cole-Parmer Buret Clamp, Three Prongs, Dual Adjustment, Medium
0802202
₹8,400.00INR / Each
+
Cole-Parmer Replacement Sleeves, Red Vinyl, Medium, 3
0802786
₹1,900.00INR / Each
=
Total Price 10300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
  • 1 year warranty
More About this Item
Used to hold apparatus at a fixed 90° angle near the lab frame where no adjustment is required after set-up. Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Utilizing thumbscrews, both prongs are easily adjusted over a wider range. Offered in either stainless steel or nickel plated zinc. Supplied with non-slip vinyl sleeves or for temperatures above 212°F (100°C), fiberglass covers.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc with red vinyl and black fiberglass sleeves
  • Clamp Type3-Prong Dual Adjust Nickel-Plated Zinc
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)2.72
  • Accepts Rod Diameter (in)0.75
  • Length (in)5.75
  • DescriptionBuret Clamp, Single, Three-Prong, Medium