Cole-Parmer Medium 2 Prong Fixed Position Clamp Stainless Steel
Representative image only.
Cole-Parmer Medium 2 Prong Fixed Position Clamp Stainless Steel
Cole-Parmer | Item: 0812996
Clamps include a built-in holder that grips rods up to ¾” (19mm)
₹42,400.00INR / Each
₹42,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Medium 2 Prong Fixed Position Clamp Stainless Steel
Cole-Parmer Medium 2 Prong Fixed Position Clamp Stainless Steel
0812996
₹42,400.00INR / Each
+
Cole-Parmer Replacement Sleeves, Red Vinyl, Medium, 2
0802780
₹1,900.00INR / Each
=
Total Price 44300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
  • Durable stainless steel clamps offer chemical resistance and increased strength
  • 2 year warranty
More About this Item
Used to hold apparatus at a fixed 90° angle near the lab frame where no adjustment is required after set-up. Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Utilizing thumbscrews, both prongs are easily adjusted over a wider range. Offered in either stainless steel or nickel plated zinc. Supplied with non-slip vinyl sleeves or for temperatures above 212°F (100°C), fiberglass covers.
Two year warranty
Specifications
  • Clamp MaterialStainless Steel
  • Clamp Type2-Prong Single Adjust Stainless Steel
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)3.03
  • Accepts Rod Diameter (in)0.75
  • Length (in)5.24
  • DescriptionMedium 2 Prong Fixed Position Clamp Stainless Steel