Cole-Parmer Replacement Sleeves, Red Vinyl, Medium, 2
Representative image only.
Cole-Parmer Replacement Sleeves, Red Vinyl, Medium, 2
Cole-Parmer | Item: 0802780
Securely hold your flasks, beakers and burettes
₹1,900.00INR / Each
₹1,900.00INR / Each
Product Description
More About this Item
Sleeves are available in both vinyl and fiberglass can be easily removed from a clamp for cleaning or replacement. Fiberglass sleeves are recommended for all applications above 212¢ªF (100¢ªC).
Specifications
  • DescriptionReplacement Sleeves, Red Vinyl, Medium, 2