Cole-Parmer® Economical Polypropylene Graduated Cylinder
Polypropylene cylinders offer very good chemical resistance
  • Meet ISO Standard 6706
  • Calibrated “to contain” for measuring final volumes
  • Molded graduations for durability
  • Autoclavable
These polypropylene (PP) graduated cylinders meet stringent international quality criteria according to ISO 6706 standards. The cylinders are chemically resistant to most acids, bases and many common solvents, and are shatter proof. They can withstand a maximum temperature of 121 °C (249.8 °F).
1,600.00 - ₹10,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
10 0.2 Polypropylene
₹1,600.00INR / Each
COMPARE
25 0.5 Polypropylene
₹1,600.00INR / Each
COMPARE
50 1 Polypropylene
₹2,100.00INR / Each
COMPARE
100 1 Polypropylene
₹2,400.00INR / Each
COMPARE
250 2 Polypropylene
₹3,700.00INR / Each
COMPARE
500 5 Polypropylene
₹5,000.00INR / Each
COMPARE
1000 10 Polypropylene
₹7,000.00INR / Each
COMPARE
2000 20 Polypropylene
₹10,000.00INR / Each
Showing 1- 8 of 8