Cole-Parmer® Dispensers for Parafilm® M Wrap
Safely store, dispense, and cut Parafilm® M tape and labels
  • Easy application with no mess
  • Holds one or two 50 mm (2”) or one 100 mm (4”) Parafilm M rolls
These Parafilm M tape dispensers feature built-in safety blade slides which allow you to cut with no danger to fingertips. Eliminates the need for scissors. Constructed of easy to clean ABS plastic.
The Parafilm M wrap (model 06720-40) is sold separately.
8,400.00 - ₹10,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Width (in)
Availability Pricing
COMPARE
Natural 4 29/32 6 51/64
₹10,400.00INR / Each
COMPARE
Blue 4 45/64 6 1/10
₹8,400.00INR / Each