Cole-Parmer® Cryovials
Reliable long term storage of your precious samples
  • Temperature range: -196°C to 121°C
  • Available in 4 different volumes: 0.5ml, 1.5ml, 2.0ml, and 5.0ml
  • Tight screw cap closures with silicone gaskets prevents liquid leakage
  • The maximum reservoir capacity is 80% of the total volume
  • Certified DNase-, RNase-free and non-pyogenic
13,000.00 - ₹73,400.00INR / Case of 1
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Sterile Tube Bottom Shape
Availability Pricing
COMPARE
0.5 No Conical
₹13,000.00INR / Case of 1
COMPARE
0.5 Yes Conical
₹17,100.00INR / Case of 1
COMPARE
1.5 No Conical
₹15,500.00INR / Case of 1
COMPARE
1.5 Yes Conical
₹33,600.00INR / Case of 1
COMPARE
2 No Conical
₹14,900.00INR / Case of 1
COMPARE
2 Yes Conical
₹32,600.00INR / Case of 1
COMPARE
0.5 No Self-standing
₹13,000.00INR / Case of 1
COMPARE
0.5 Yes Self-standing
₹25,200.00INR / Case of 1
COMPARE
1.5 No Self-standing
₹14,400.00INR / Case of 1
COMPARE
1.5 Yes Self-standing
₹17,000.00INR / Case of 1
COMPARE
2 No Self-standing
₹16,200.00INR / Case of 1
COMPARE
2 Yes Self-standing
₹18,400.00INR / Case of 1
COMPARE
0.5 No Conical
₹53,100.00INR / Case of 1
COMPARE
1.5 No Conical
₹61,500.00INR / Case of 1
COMPARE
2 No Conical
₹73,400.00INR / Case of 1
COMPARE
0.5 No Self-standing
₹53,100.00INR / Case of 1
COMPARE
1.5 No Self-standing
₹61,500.00INR / Case of 1
COMPARE
2 No Self-standing
₹67,700.00INR / Case of 1
COMPARE
5 Yes Self-standing
₹19,100.00INR / Case of 1
Showing 1- 19 of 19