Cole-Parmer® Clear Disposable Syringes
Luer-lock or slip tips allow for easy connection of syringe filters
  • Constructed of durable polypropylene
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-lock or Luer-slip tip connections. Luer-lock syringes have a twist-and-lock mechanism to create a secure fitting connection, making them a safe choice for high back pressure liquids in filter applications. Luer-slip syringes slide into place and are held by friction—ideal for low-pressure applications.
Syringes come non-sterilized in bulk bags or packages. Model 07945-43 comes sterile. Needles and shield tips are not included.
6,100.00 - ₹17,000.00INR / Bag of 200
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Syringe Capacity (mL) Syringe Type
Availability Pricing
COMPARE
No 1 Luer slip tip
₹17,000.00INR / Bag of 450
COMPARE
No 3 Luer lock tip
₹12,100.00INR / Bag of 350
COMPARE
No 3 Luer centric slip tip
₹12,500.00INR / Bag of 350
COMPARE
No 5 Luer lock tip
₹7,500.00INR / Bag of 200
COMPARE
No 5 Luer centric slip tip
₹7,900.00INR / Bag of 200
COMPARE
No 10 Luer lock tip
₹7,900.00INR / Bag of 130
COMPARE
No 10 Luer centric slip tip
₹7,900.00INR / Bag of 130
COMPARE
No 20 Luer lock tip
₹6,100.00INR / Bag of 50
COMPARE
No 20 Luer eccentric slip tip
₹8,000.00INR / Bag of 50
COMPARE
No 30 Luer lock tip
₹7,500.00INR / Bag of 50
COMPARE
No 30 Luer eccentric slip tip
₹7,800.00INR / Bag of 50
COMPARE
No 60 Luer lock tip
₹7,600.00INR / Bag of 25
COMPARE
Yes 60 Luer slip tip
₹7,000.00INR / Box of 25
Showing 1- 13 of 13