Cole-Parmer® Cleanroom Female Luer Fittings, Straight Union
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Forms a leak-free luer connection with a quick simple turn
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
These Cole-Parmer® cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. This straight union features a female luer coupler on each end to connect two male luer fittings together. Unions are available in five different formulations—select one that best suits your application.
950.00 - ₹1,700.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material
Availability Pricing
COMPARE
Polycarbonate
₹950.00INR / Pkg of 10
COMPARE
Nylon
₹1,400.00INR / Pkg of 10
COMPARE
PVDF
₹1,700.00INR / Pkg of 10
COMPARE
Nylon
₹990.00INR / Pkg of 10
COMPARE
Polypropylene
₹1,400.00INR / Pkg of 10