Cole-Parmer Luer Fitting, Straight Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Female Luer; 10/Pk
Representative image only.
Cole-Parmer Luer Fitting, Straight Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Female Luer; 10/Pk
Cole-Parmer | Item: 5011425
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹1,400.00INR / Pkg of 10
₹1,400.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Luer Fitting, Straight Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Female Luer; 10/Pk
Cole-Parmer Luer Fitting, Straight Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Female Luer; 10/Pk
5011425
₹1,400.00INR / Pkg of 10
+
Tygon E-3603 Tubing, 3/16" ID x 5/16" OD; 50 Ft
5010628
Call For Price
+ + =
Total Price 5000.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Forms a leak-free luer connection with a quick simple turn
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Animal-derivative-free polypropylene
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
More About this Item
These Cole-Parmer® cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. This straight union features a female luer coupler on each end to connect two male luer fittings together. Unions are available in five different formulations—select one that best suits your application.
Animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolypropylene
  • Number Of Ports2
  • Regulatory ApprovalsADCF;FDA;Non-Phthalate;RoHS;Non-DEHP
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Female Luer
  • DescriptionLuer Fitting, Straight Union, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Female Luer; 10/Pk