Cole-Parmer Luer Fitting, Straight Union, Natural Nylon, Cleanroom Packed, Female Luer; 10/Pk
Representative image only.
Cole-Parmer Luer Fitting, Straight Union, Natural Nylon, Cleanroom Packed, Female Luer; 10/Pk
Cole-Parmer | Item: 5011424
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹990.00INR / Pkg of 10
₹990.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Luer Fitting, Straight Union, Natural Nylon, Cleanroom Packed, Female Luer; 10/Pk
Cole-Parmer Luer Fitting, Straight Union, Natural Nylon, Cleanroom Packed, Female Luer; 10/Pk
5011424
₹990.00INR / Pkg of 10
+
Tygon E-3603 Tubing, 1/16" ID x 3/16" OD; 50 Ft
5010622
Call For Price
+ =
Total Price 1390.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Forms a leak-free luer connection with a quick simple turn
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Natural nylon
  • RoHS compliant
More About this Item
These Cole-Parmer® cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. This straight union features a female luer coupler on each end to connect two male luer fittings together. Unions are available in five different formulations—select one that best suits your application.
Natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Number Of Ports2
  • Regulatory ApprovalsNon-Phthalate;Non-DEHP;RoHS
  • Luer Connection Type (Port 1)Female Luer
  • Luer Connection Type (Port 2)Female Luer
  • DescriptionLuer Fitting, Straight Union, Natural Nylon, Cleanroom Packed, Female Luer; 10/Pk