Cole-Parmer® Broad-Spout PTFE Beakers
Stand up to the harshest chemicals
  • Pour easily with broad spout
  • Autoclavable
These beakers are constructed of PTFE making them ideal for use with most harsh chemicals. The broad spout allows easy pouring, and these beakers can withstand temperatures from -450 to 550°F (-268 to 288°C).
Beakers can be used with caustic chemicals and when purity is desired.
6,300.00 - ₹175,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Outside Diameter (OD) (in)
Availability Pricing
COMPARE
1 Polytetrafluoroethylene (PTFE) 1/2
₹7,600.00INR / Pkg of 2
COMPARE
5 Polytetrafluoroethylene (PTFE) 3/4
₹9,200.00INR / Pkg of 2
COMPARE
10 Polytetrafluoroethylene (PTFE) 1
₹9,900.00INR / Pkg of 2
COMPARE
25 Polytetrafluoroethylene (PTFE) 1 1/4
₹14,700.00INR / Pkg of 2
COMPARE
50 Polytetrafluoroethylene (PTFE) 1 3/4
₹8,400.00INR / Each
COMPARE
100 Polytetrafluoroethylene (PTFE) 2 1/8
₹6,300.00INR / Each
COMPARE
150 Polytetrafluoroethylene (PTFE) 2 3/8
₹11,900.00INR / Each
COMPARE
250 Polytetrafluoroethylene (PTFE) 2 5/8
₹12,700.00INR / Each
COMPARE
400 Polytetrafluoroethylene (PTFE) 3 1/8
₹23,700.00INR / Each
COMPARE
500 Polytetrafluoroethylene (PTFE) 3 1/8
₹23,700.00INR / Each
COMPARE
600 Polytetrafluoroethylene (PTFE) 3 1/2
₹32,100.00INR / Each
COMPARE
1000 Polytetrafluoroethylene (PTFE) 4
₹51,900.00INR / Each
COMPARE
2000 Polytetrafluoroethylene (PTFE) 4 3/4
₹82,200.00INR / Each
COMPARE
5000 Polytetrafluoroethylene (PTFE) 6 3/4
₹175,000.00INR / Each
Showing 1- 14 of 14