Cole-Parmer® Adjustable-Volume Pipettor, Tips, and Reagent Reservoir Bundle
Ergonomic finger grip for comfortable use
  • Bundle combines pipettor, tips, and reservoir for added convenience
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Adjustable-volume pipettors feature click-stop volume settings
  • Leach-free tips ensure maximum recovery of sample yield
Bundled for convenience, the adjustable-volume pipettor, pipette tips, and reagent reservoirs provide you with everything you need to keep dispensing consistent and reliable. The pipettor is easy to operate, with an ergonomic, light-weight push-button design and volume displayed through handgrip window for quick glance convenience.
The single channel reagent reservoirs feature a V-shaped bottom designed to maximize reagent retrieval and reduce waste. Reservoirs are constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring.
26,600.00 - ₹27,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of Sterile
Availability Pricing
COMPARE
1000 Tips: Standard 6 x 96 Racks; Reservoirs: Individually Wrapped 50/Case Yes
₹27,900.00INR / Each
COMPARE
1000 Tips: Bulk pack 1000/Bg; Reservoirs: Bulk pack 100/Cs No
₹26,600.00INR / Each
COMPARE
5000 Tips: Bulk pack 100/Bg; Reservoirs: Bulk pack 100/Cs No
₹27,900.00INR / Each