Cole-Parmer Adjustable-Volume Pipettor Bundle, 500 to 5000 µL Pipetor and Bulk Pipette Tips, 25 mL Bulk Packed Reagent Reservoirs; 100 Reservoirs, 100 Tips
Representative image only.
Cole-Parmer Adjustable-Volume Pipettor Bundle, 500 to 5000 µL Pipetor and Bulk Pipette Tips, 25 mL Bulk Packed Reagent Reservoirs; 100 Reservoirs, 100 Tips
Cole-Parmer | Item: 2530707
Ergonomic finger grip for comfortable use
₹27,900.00INR / Each
₹27,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Adjustable-Volume Pipettor Bundle, 500 to 5000 µL Pipetor and Bulk Pipette Tips, 25 mL Bulk Packed Reagent Reservoirs; 100 Reservoirs, 100 Tips
Cole-Parmer Adjustable-Volume Pipettor Bundle, 500 to 5000 µL Pipetor and Bulk Pipette Tips, 25 mL Bulk Packed Reagent Reservoirs; 100 Reservoirs, 100 Tips
2530707
₹27,900.00INR / Each
+ =
Total Price 38200.00 ADD 2 ITEMS TO CART
Product Description
Includes
one adjustable-volume pipettor, 100 bulk packed 500 to 5000 µL pipette tips, and 100 single channel 25 mL reagent reservoirs.
Key Features
  • Bundle combines pipettor, tips, and reservoir for added convenience
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Adjustable-volume pipettors feature click-stop volume settings
  • Leach-free tips ensure maximum recovery of sample yield
  • Dispensing Volume of 500 to 5000 µL
  • Reagent reservoirs and pipette tips are not sterile and bulk packed
  • 3 year warranty
More About this Item
Bundled for convenience, the adjustable-volume pipettor, pipette tips, and reagent reservoirs provide you with everything you need to keep dispensing consistent and reliable. The pipettor is easy to operate, with an ergonomic, light-weight push-button design and volume displayed through handgrip window for quick glance convenience.
The single channel reagent reservoirs feature a V-shaped bottom designed to maximize reagent retrieval and reduce waste. Reservoirs are constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
3 Year WarrantyConformite Europeenne
Specifications
  • SterileNo
  • Max Volume (µL)5000
  • Min Volume (µL)500
  • Increments (µL)50
  • Accuracy (%)0.5
  • Number Of Channels1
  • Tip StyleStandard, 12.06 cm L
  • Tip EjectorYes
  • Wetted MatterialPolypropylene tips and polystyrene reservoirs
  • Color CodeGreen
  • Filter TypeNone
  • Package OfTips: Bulk pack 100/Bg; Reservoirs: Bulk pack 100/Cs
  • DescriptionAdjustable-Volume Pipettor Bundle, 500 to 5000 µL Pipetor and Bulk Pipette Tips, 25 mL Bulk Packed Reagent Reservoirs; 100 Reservoirs, 100 Tips
  • Warranty3 Years