Cole-Parmer Adjustable-Volume Pipettor Bundle, 100 to 1000 µL Pipetor and Sterile Pipette Tips, 25 mL Sterile Individually Wrapped Reagent Reservoirs; 50 Reservoirs, 6 x 96 Tip Racks
Representative image only.
Cole-Parmer Adjustable-Volume Pipettor Bundle, 100 to 1000 µL Pipetor and Sterile Pipette Tips, 25 mL Sterile Individually Wrapped Reagent Reservoirs; 50 Reservoirs, 6 x 96 Tip Racks
Cole-Parmer | Item: 2530705
Ergonomic finger grip for comfortable use
₹27,900.00INR / Each
₹27,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Adjustable-Volume Pipettor Bundle, 100 to 1000 µL Pipetor and Sterile Pipette Tips, 25 mL Sterile Individually Wrapped Reagent Reservoirs; 50 Reservoirs, 6 x 96 Tip Racks
Cole-Parmer Adjustable-Volume Pipettor Bundle, 100 to 1000 µL Pipetor and Sterile Pipette Tips, 25 mL Sterile Individually Wrapped Reagent Reservoirs; 50 Reservoirs, 6 x 96 Tip Racks
2530705
₹27,900.00INR / Each
+ =
Total Price 36500.00 ADD 2 ITEMS TO CART
Product Description
Includes
one adjustable-volume pipettor, six racks of 96 sterile 100 to 1000 µL pipette tips, and 50 single channel 25 mL sterile reagent reservoirs.
Key Features
  • Bundle combines pipettor, tips, and reservoir for added convenience
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
  • Adjustable-volume pipettors feature click-stop volume settings
  • Leach-free tips ensure maximum recovery of sample yield
  • Dispensing Volume of 100 to 1000 µL
  • Reagent reservoirs are sterile and individually wrapped to reduce cross contamination
  • Sterile pipette tips are packaged in 6 standard racks of 96 tips each
  • 3 year warranty
More About this Item
Bundled for convenience, the adjustable-volume pipettor, pipette tips, and reagent reservoirs provide you with everything you need to keep dispensing consistent and reliable. The pipettor is easy to operate, with an ergonomic, light-weight push-button design and volume displayed through handgrip window for quick glance convenience.
The single channel reagent reservoirs feature a V-shaped bottom designed to maximize reagent retrieval and reduce waste. Reservoirs are constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
3 Year WarrantyConformite Europeenne
Specifications
  • SterileYes
  • Max Volume (µL)1000
  • Min Volume (µL)100
  • Increments (µL)5
  • Accuracy (%)0.6
  • Number Of Channels1
  • Tip StyleStandard, 8.35 cm L
  • Tip EjectorYes
  • Wetted MatterialPolypropylene tips and polystyrene reservoirs
  • Color CodeBlue
  • Filter TypeNone
  • Package OfTips: Standard 6 x 96 Racks; Reservoirs: Individually Wrapped 50/Case
  • DescriptionAdjustable-Volume Pipettor Bundle, 100 to 1000 µL Pipetor and Sterile Pipette Tips, 25 mL Sterile Individually Wrapped Reagent Reservoirs; 50 Reservoirs, 6 x 96 Tip Racks
  • Warranty3 Years