Cole-Parmer® 96-Well Deep Well Plates
Meets SBS guidelines for robotic handling
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Autoclavable at 121°C for 20 minutes (polypropylene plates only)
  • Alphanumeric pattern for simple sample identification
The 96-well deep well plates feature rounded bottoms, are compatible with most robotic samplers, and available in 0.5 mL, 1.2 mL (1 mL capped), and 2.2 mL (2 mL capped) well volumes.
39,900.00 - ₹104,000.00INR / Case of 56
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Well Shape Well Volume (mL)
Availability Pricing
COMPARE
No Round 0.5
₹91,800.00INR / Case of 56
COMPARE
Yes Round 0.5
₹104,000.00INR / Case of 56
COMPARE
No Round 1.2
₹39,900.00INR / Case of 32
COMPARE
Yes Round 1.2
₹47,100.00INR / Case of 32
COMPARE
No Round 1.2
₹39,900.00INR / Case of 32
COMPARE
Yes Round 1.2
₹50,800.00INR / Case of 32
COMPARE
No Round, raised rim 1.2
₹39,900.00INR / Case of 32
COMPARE
Yes Round, raised rim 1.2
₹47,100.00INR / Case of 32
COMPARE
No Round, raised rim 1.2
₹39,900.00INR / Case of 32
COMPARE
Yes Round, raised rim 1.2
₹47,100.00INR / Case of 32
COMPARE
No Square, round bottom 2.2
₹63,700.00INR / Case of 32
COMPARE
Yes Square, round bottom 2.2
₹63,700.00INR / Case of 32
Showing 1- 12 of 12