Chemical Indicators, ProChem BD Test Paper
Test papers are useful for daily monitoring of pre-vacuum steam sterilizers to detect air leaks,
inadequate steam penetration and vacuum pump failures. The colour change is from light blue to a
uniform black colour for the test paper and from pink to a uniform black colour for the test pack.
Meets AAMI ST66 and EN867-3 test methods for detecting the presence of air.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Capacity (cu ft) Max Pressure (PSI) Max Temperature (° F)
Availability Pricing
ProChem BDTPA4 BD Test Paper - Class 3 (8" X 10"), 25 Papers/pack WW-94702-22 Mfr # BDTPA4
COMPARE
Call For Price
ProChem BDTPSQ BD Test Pack - Class 3, 5 Pkts/box WW-94702-23 Mfr # BDTPSQ