ProChem BDTPSQ BD Test Pack - Class 3, 5 Pkts/box
Representative image only.
ProChem BDTPSQ BD Test Pack - Class 3, 5 Pkts/box
ProChem | Mfr: BDTPSQ| Item: 9470223
Call For Price
Call For Price
Product Description
More About this Item
Test papers are useful for daily monitoring of pre-vacuum steam sterilizers to detect air leaks,
inadequate steam penetration and vacuum pump failures. The colour change is from light blue to a
uniform black colour for the test paper and from pink to a uniform black colour for the test pack.
Meets AAMI ST66 and EN867-3 test methods for detecting the presence of air.
Specifications
  • DescriptionBD Test Pack - Class 3, 5 Pkts/box