CELLTREAT Scientific Products Bio-Reaction Centrifuge Tubes
Convenient and economical option for small scale, high density suspension cell culture
  • Feature a vented cap with a 0.22 μm hydrophobic membrane—ideal for biological reaction experiments that require aseptic conditions with free gas exchange
  • Black graduations make reading liquid-levels easy
  • Large, white marking area simplifies labeling
39,500.00 - ₹120,000.00INR / Case of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Packaging Tube Bottom Shape Tube Capacity (mL)
Availability Pricing
CELLTREAT Scientific Products 229471 Bio-Reaction Centrifuge Tube, 15 mL, Vented Cap, Bag, Sterile; 100/Cs 0633640 Mfr # 229471
COMPARE
10 per resealable bag; 10 bags per case Conical 15
₹39,500.00INR / Case of 100
CELLTREAT Scientific Products 229472 Bio-Reaction Centrifuge Tube, 15 mL, Vented Cap, Foam Rack, Sterile; 300/Cs 0633641 Mfr # 229472
COMPARE
50 per foam rack; 6 racks per case Conical 15
₹110,000.00INR / Case of 300
CELLTREAT Scientific Products 229475 Bio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Bag, Sterile; 100/Cs 0633643 Mfr # 229475
COMPARE
10 per resealable bag; 10 bags per case Conical 50
₹46,200.00INR / Case of 100
CELLTREAT Scientific Products 229476 Bio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Foam Rack, Sterile; 300/Cs 0633644 Mfr # 229476
COMPARE
25 per foam rack; 12 racks per case Conical 50
₹120,000.00INR / Case of 300
CELLTREAT Scientific Products 229479 Bio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Standing, Bag, Sterile; 100/Cs 0633647 Mfr # 229479
COMPARE
10 per resealable bag; 10 bags per case Conical/Skirted 50
₹52,700.00INR / Case of 100