CELLTREAT Scientific Products 229471 Bio-Reaction Centrifuge Tube, 15 mL, Vented Cap, Bag, Sterile; 100/Cs
Representative image only.
CELLTREAT Scientific Products 229471 Bio-Reaction Centrifuge Tube, 15 mL, Vented Cap, Bag, Sterile; 100/Cs
CELLTREAT Scientific Products | Mfr: 229471| Item: 0633640
Convenient and economical option for small scale, high density suspension cell culture
₹39,500.00INR / Case of 100
₹39,500.00INR / Case of 100
Buy It Together
Current Item
CELLTREAT Scientific Products 229471 Bio-Reaction Centrifuge Tube, 15 mL, Vented Cap, Bag, Sterile; 100/Cs
CELLTREAT Scientific Products 229471 Bio-Reaction Centrifuge Tube, 15 mL, Vented Cap, Bag, Sterile; 100/Cs
0633640
₹39,500.00INR / Case of 100
+ + =
Total Price 89500.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Feature a vented cap with a 0.22 μm hydrophobic membrane—ideal for biological reaction experiments that require aseptic conditions with free gas exchange
  • Black graduations make reading liquid-levels easy
  • Large, white marking area simplifies labeling
Sterile
Specifications
  • Max RCF x g12000
  • SterileYes
  • Tube Capacity (mL)15
  • Tube MaterialPolypropylene
  • Tube ColorClear
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleVented
  • GraduationsYes
  • Packaging10 per resealable bag; 10 bags per case
  • Special FeaturesSterile; bagged
  • DescriptionBio-Reaction Centrifuge Tube, 15 mL, Vented Cap, Bag, Sterile; 100/Cs