CELLTREAT Scientific Products 229476 Bio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Foam Rack, Sterile; 300/Cs
Representative image only.
CELLTREAT Scientific Products 229476 Bio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Foam Rack, Sterile; 300/Cs
CELLTREAT Scientific Products | Mfr: 229476| Item: 0633644
Convenient and economical option for small scale, high density suspension cell culture
₹120,000.00INR / Case of 300
₹120,000.00INR / Case of 300
Buy It Together
Current Item
CELLTREAT Scientific Products 229476 Bio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Foam Rack, Sterile; 300/Cs
CELLTREAT Scientific Products 229476 Bio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Foam Rack, Sterile; 300/Cs
0633644
₹120,000.00INR / Case of 300
+ + =
Total Price 173100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Feature a vented cap with a 0.22 μm hydrophobic membrane—ideal for biological reaction experiments that require aseptic conditions with free gas exchange
  • Black graduations make reading liquid-levels easy
  • Large, white marking area simplifies labeling
Sterile
Specifications
  • Max RCF x g16000
  • SterileYes
  • Tube Capacity (mL)50
  • Tube MaterialPolypropylene
  • Tube ColorClear
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap StyleVented
  • GraduationsYes
  • Packaging25 per foam rack; 12 racks per case
  • Special FeaturesSterile; foam rack
  • DescriptionBio-Reaction Centrifuge Tube, 50 mL, Vented Cap, Foam Rack, Sterile; 300/Cs