CELLTREAT Multiwell Tissue Culture Plates
Alphanumeric markings for easy identification of individual wells
Raised rim on each well nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross-contamination. Stacking ring on the lid offers added stability when handling tall stacks of plates. Clear polystyrene construction lets you easily view samples. Plates are sterilized by gamma irradiation.
23,900.00 - ₹55,400.00INR / Case of 100
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Number of Wells Surface Treatment Well Shape
Availability Pricing
CELLTREAT Scientific Products 229512 12-Well Cell Culture Plate with Lid; 100/cs WW-01931-13 Mfr # 229512
COMPARE
12 Flat
₹46,500.00INR / Case of 100
CELLTREAT Scientific Products 229524 24-Well Cell Culture Plate with Lid; 100/cs WW-01931-15 Mfr # 229524
COMPARE
24 Flat
₹47,700.00INR / Case of 100
CELLTREAT Scientific Products 229105 6-Well Treated Culture Plate with Lid; 50/cs WW-01959-01 Mfr # 229105
COMPARE
6 Yes Flat
₹24,200.00INR / Case of 50
CELLTREAT Scientific Products 229111 12-Well Treated Culture Plate with Lid; 50/cs WW-01959-03 Mfr # 229111
COMPARE
12 Yes Flat
₹23,900.00INR / Case of 50
CELLTREAT Scientific Products 229123 24-Well Treated Cell Culture Plate with Lid; 50/cs WW-01959-05 Mfr # 229123
COMPARE
24 Yes Flat
₹24,400.00INR / Case of 50
CELLTREAT Scientific Products 229147 48-Well Treated Cell Culture Plate with Lid; 50/cs WW-01959-07 Mfr # 229147
COMPARE
48 Yes Flat
₹26,900.00INR / Case of 50
CELLTREAT Scientific Products 229190 96-Well Treated Cell Culture Plate with Lid; 100/cs WW-01959-09 Mfr # 229190
COMPARE
96 Yes Round
₹53,500.00INR / Case of 100
CELLTREAT Scientific Products 229195 96-Well Treated Cell Culture Plate with Lid; 50/cs WW-01959-11 Mfr # 229195
COMPARE
96 Yes Flat
₹27,700.00INR / Case of 50
CELLTREAT Scientific Products 229197 96-Well Treated Cell Culture Plate with Lid; 100/cs WW-01959-13 Mfr # 229197
COMPARE
96 Yes Flat
₹46,500.00INR / Case of 100
CELLTREAT Scientific Products 229548 48-Well Cell Culture Plate with Lid; 100/cs WW-01959-15 Mfr # 229548
COMPARE
48 Flat
₹48,600.00INR / Case of 100
CELLTREAT Scientific Products 229590 96-Well Cell Culture Plate with Lid; 100/cs WW-01959-21 Mfr # 229590
COMPARE
96 Round
₹55,400.00INR / Case of 100
CELLTREAT Scientific Products 229591 96-Well Cell Culture Plate; 100/cs WW-01959-23 Mfr # 229591
COMPARE
96 Round
₹48,900.00INR / Case of 100
CELLTREAT Scientific Products 229592 96-Well Cell Culture Plate; 100/cs WW-01959-25 Mfr # 229592
COMPARE
96 Flat
₹46,800.00INR / Case of 100
CELLTREAT Scientific Products 229596 96-Well Cell Culture Plate with Lid; 100/cs WW-01959-27 Mfr # 229596
COMPARE
96 Flat
₹50,300.00INR / Case of 100
CELLTREAT Scientific Products 229506 6-Well Cell Culture Plate with Lid; 100/cs WW-01959-29 Mfr # 229506
COMPARE
6 Flat
₹45,300.00INR / Case of 100
Showing 1- 15 of 15