CELLTREAT Scientific Products 229123 24-Well Treated Cell Culture Plate with Lid; 50/cs
Representative image only.
CELLTREAT Scientific Products 229123 24-Well Treated Cell Culture Plate with Lid; 50/cs
CELLTREAT Scientific Products | Mfr: 229123| Item: 0195905
Alphanumeric markings for easy identification of individual wells
₹32,700.00INR / Case of 50
₹32,700.00INR / Case of 50
Product Description
Includes
Lids.
Key Features
  • Treated tissue culture plates have a hydrophilic surface that is ideal for cell attachment
More About this Item
Raised rim on each well nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross-contamination. Stacking ring on the lid offers added stability when handling tall stacks of plates. Clear polystyrene construction lets you easily view samples. Plates are sterilized by gamma irradiation.
Plates are sterilized by gamma irradiation and individually packaged.
Sterile
Specifications
  • Number of Wells24
  • Well ShapeFlat
  • Culture Area (cm²)1.93
  • Surface TreatmentYes
  • Lid (Included)Yes
  • SterileYes
  • Description24-Well Treated Cell Culture Plate with Lid; 50/cs