CELLTREAT Scientific Products 229596 96-Well Cell Culture Plate with Lid; 100/cs
Representative image only.
CELLTREAT Scientific Products 229596 96-Well Cell Culture Plate with Lid; 100/cs
CELLTREAT Scientific Products | Mfr: 229596| Item: 0195927
Alphanumeric markings for easy identification of individual wells
₹68,300.00INR / Case of 100
₹68,300.00INR / Case of 100
Product Description
Includes
Lids.
Key Features
  • Untreated surfaces do not facilitate cell attachment—ideal for suspensions
More About this Item
Raised rim on each well nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross-contamination. Stacking ring on the lid offers added stability when handling tall stacks of plates. Clear polystyrene construction lets you easily view samples. Plates are sterilized by gamma irradiation.
Plates are sterilized by gamma irradiation and individually packaged. 50 plates/sleeve.
Sterile
Specifications
  • Number of Wells96
  • Min Working Well Volume (mL)0.05
  • Max Working Well Volume (mL)0.3
  • Well ShapeFlat
  • Lid (Included)Yes
  • SterileYes
  • Description96-Well Cell Culture Plate with Lid; 100/cs