Cecomp® Test + Battery-Powered Digital Pressure Gauges
Introducing a precision-engineered test gauge with a standout ±0.1% accuracy for unparalleled reliability in diverse applications
  • Exceptional accuracy with ±0.015% full-scale sensor hysteresis and ±0.01% full-scale sensor repeatability
  • Versatile 4-digit LCD display, offering three readings per second and a configurable white LED backlight
  • Rugged design meets NEMA 2 (IP51) protection for durability in challenging environments
  • User-friendly controls include a three-button keypad with passcode-protected calibration, engineering unit selection, configurable min/max memory, and auto shutoff times
These precision-engineered test gauges stand out with exceptional ±0.1% full scale accuracy, offering reliability across diverse applications. With a versatile display featuring a 4-digit LCD and configurable white LED backlight, it provides real-time readings and user-friendly controls through a three-button keypad. The ruggedized design ensures durability with 316L stainless steel wetted parts, an epoxy powder-coated aluminum case with case stiffener, ABS/polycarbonate bezel, coated circuit boards for humidity resistance, front and rear rubber gaskets, polycarbonate label, and a protective red rubber boot. User-configurable options, such as selectable units, auto shutoff times, and min/max memory, enhance customization and convenience. Additionally, the gauge's comprehensive features include wide operational temperatures, overpressure protection of 5000 psig for 3000 psig sensors or 7500 psig for 5000 psig sensors, burst pressure resilience of 4x the sensor pressure rating*, and vacuum service capability, making these versatile and robust tools for various industrial and environmental conditions.
122,000.00INR / Each
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
Compareitem specifications
Accuracy Process Connection Description
Availability Pricing
Cecomp Test + Battery-Powered Digital Vacuum Gauge, 0 to 14.70 psig 6893700 Mfr # CTP3B15PSIVAC
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Vacuum Gauge, 0 to 14.70 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Absolute Reference Gauge, 15.00 to 0 psi 6893701 Mfr # CTP3B15PSIA
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Absolute Reference Gauge, 15.00 to 0 psi
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Absolute Reference Gauge, 30.00 to 0 psi 6893702 Mfr # CTP3B30PSIA
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Absolute Reference Gauge, 30.00 to 0 psi
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Absolute Reference Gauge, 100.0 to 0 psi 6893703 Mfr # CTP3B100PSIA
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Absolute Reference Gauge, 100.0 to 0 psi
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Vacuum/Pressure Gauge, -14.70 to 15.00 psig 6893704 Mfr # CTP3B+/-15PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Vacuum/Pressure Gauge, -14.70 to 15.00 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Vacuum/Pressure Gauge, -29.92 inHg to 15.00 psig 6893705 Mfr # CTP3B30V15PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Vacuum/Pressure Gauge, -29.92 inHg to 15.00 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Vacuum/Pressure Gauge, -14.7 to 100.0 psig 6893706 Mfr # CTP3B15V100PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Vacuum/Pressure Gauge, -14.7 to 100.0 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Vacuum/Pressure Gauge, -29.9 inHg to 100.0 psig 6893707 Mfr # CTP3B30V100PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Vacuum/Pressure Gauge, -29.9 inHg to 100.0 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Vacuum/Pressure Gauge, -14.7 to 200.0 psig 6893708 Mfr # CTP3B15V200PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Vacuum/Pressure Gauge, -14.7 to 200.0 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Vacuum/Pressure Gauge, -29.92 in Hg to 200.0 psig 6893709 Mfr # CTP3B30V200PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Vacuum/Pressure Gauge, -29.92 in Hg to 200.0 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 3.000 psig 6893710 Mfr # CTP3B3PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 3.000 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 5.000 psig 6893711 Mfr # CTP3B5PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 5.000 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 15.000 psig 6893712 Mfr # CTP3B15PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 15.000 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 30.00 psig 6893713 Mfr # CTP3B30PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 30.00 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 60.00 psig 6893714 Mfr # CTP3B60PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 60.00 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 100.0 psig 6893715 Mfr # CTP3B100PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 100.0 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 200.0 psig 6893716 Mfr # CTP3B200PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 200.0 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 500.0 psig 6893717 Mfr # CTP3B500PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 500.0 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 1000 psig 6893718 Mfr # CTP3B1000PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 1000 psig
₹122,000.00INR / Each
Cecomp Test + Battery-Powered Digital Pressure Gauge, 0 to 2000 psig 6893719 Mfr # CTP3B2000PSIG
COMPARE
±0.1% full-scale 1/4" NPT-male Battery-Powered Digital Pressure Gauge, 0 to 2000 psig
₹122,000.00INR / Each
Showing 1 - 20 of 22