Bytac Surface Protectors
Prevents corrosion and absorption of toxic chemicals
  • Easy-to-apply to most work surfaces
  • Easy-to-clean
  • Non-contaminating
  • Cuts easily with scissors
Bytac surface protectors are pressure sensitive FEP film that is inert to most chemicals. These easy-to-use adhesive surface protectors are ideal for benchtops, tables, fume hoods, and work surfaces. Use to prevent corrosion and absorption of toxic or radioactive chemicals. The FEP film comes with a vinyl or aluminum backing. Choose vinyl backing for temperatures up to 200°>F (93°>C). Aluminum backing withstands temperatures up to 360°>F (182°>C) and provides a nonstick surface when used in ovens. Both types of surface protectors are easy to cut. Pressure-sensitive adhesive holds surface protectors in place—just remove paper backing and apply. Five yards/roll.
53,100.00 - ₹118,000.00INR / Roll of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Thickness (in) Width (in)
Availability Pricing
Bytac D1069324 Surface Protector,FEP, White, Vinyl 25" x 15' roll WW-06804-00 Mfr # D1069324 BYTAC
COMPARE
FEP 0.01 25
₹100,000.00INR / Roll of 1
Bytac D1069329 Surface Protector,FEP, White, Aluminum 25" x 15' roll WW-06804-10 Mfr # D1069329 BYTAC
COMPARE
FEP 0.0001 25
₹118,000.00INR / Roll of 1
Bytac D9860005 Surface Protector, FEP, White, Vinyl 12 1/2" x 15' roll WW-06804-20 Mfr # D9860005 BYTAC
COMPARE
FEP 0.3125 12.5
₹53,100.00INR / Roll of 1
Bytac D9860012 Surface Protector, FEP, White, Aluminum 12 1/2" x 15' roll WW-06804-30 Mfr # D9860012 BYTAC
COMPARE
FEP 0.0781 12.5
₹53,100.00INR / Roll of 1