In Stock
Thickness (mm)
Thickness (in)
Width (cm)
Width (in)
Material
Bytac
Bytac
Bytac adhesive surface protectors keep tables, benchtops, fume hoods, and other surfaces free from absorbing chemicals that may be toxic or radioactive. Choose from vinyl or aluminum backings. We are a Bytac authorized distributor.