Burkle Disposable Double-Sided Sampling Spatulas
No more cross-contamination from reusing stainless steel spatulas
  • Polystyrene is food-safe and complies with EU and FDA regulations
  • Trust cleanroom class 7 production, assembly, and packaging for critical transfers
  • Available bulk-packed or individually wrapped and gamma sterilized
Double-sided spatula and micro spatula features rounded ends and a sturdy center handle. Ensure precise work with either the 1.0-mL or 250-µL side, perfect for small amounts of liquids, powders, granulates, pastes, or creams.
Double-sided spoon and spatula features a 500-µL scoop and a 17-mm spatula blade. Easily measure, transfer, scrape, mix, and stir—especially well-suited for applications with both liquid and solid components.
7,500.00 - ₹18,600.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Sterile Volume (mL)
Availability Pricing
Burkle 5378-0031 Disposable double ended spoon and spatula, PS, .5 mL spoon, 17 mm blade WW-06042-49 Mfr # 5378-0031
COMPARE
Polystyrene (PS) No 0.5
₹7,500.00INR / Pkg of 100
Burkle 5378-0032 Disposable double ended micro spatula, PS; 1.0 ml and 2.5 mL WW-06042-50 Mfr # 5378-0032
COMPARE
Polystyrene (PS) No 1
₹7,500.00INR / Pkg of 100
Burkle 5378-1031 Disposable double ended spoon and spatula, PS, .5 mL spoon, 17 mm blade, sterile WW-06042-69 Mfr # 5378-1031
COMPARE
Polystyrene (PS) Yes 0.5
₹18,600.00INR / Pkg of 100
Burkle 5378-1032 Disposable double ended micro spatula, PS,sterile; 1.0 ml and 2.5 mL WW-06042-70 Mfr # 5378-1032
COMPARE
Polystyrene (PS) Yes 1
₹18,600.00INR / Pkg of 100