Bullard® Filter Cartridge for EVA PAPR

Bullard Filters are designed for use with EVA series PAPR system providing protection not only against particulates but also against harmful organic vapours and chemical gases.
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.

  • The range available in three variants
1. Bullard® PAPRFC3 High Efficiency (HE) Particulate Filter for EVA PAPR:

Bullard HEPA Filter is designed to work with the EVA series PAPR system. HEPA filter has 99.97% efficiency against solid or liquid particles including oil-based particles.

Key Features:
• Filter Type: Mechanical
• Protection: Particulates
• Efficiency: High Efficiency Particulate Absolute (HEPA) 99.99% vs. a mass mean 0.3 micron aerosol at 85 LPM
• Filter Housing Material: ABS Plastic

Compliance:
NIOSH 42 CFR Part 84, EN 12941, CE 1094

2. Bullard® PAPRFC4 OVAGE Filter for EVA PAPR:

Bullard PAPRFC4 OVAGE filter is designed for use with EVA series PAPR system providing protection not only against particulates but also against harmful organic vapours and chemical gases.

Key Features:
• Filter Type: Combination Mechanical & Chemical
• Protection: Particulates, Organic Vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, and Hydrogen Fluoride
• Particulate Efficiency: High Efficiency Particulate Absolute (HEPA) 99.99% vs. a mass mean 0.3 micron aerosol at 85 LPM Media secured with urethane glue around perimeter
• Filter Housing Material: ABS Plastic
• Sorbent Bed: Activated Carbon (Chromium Free)

Compliance:
Tested in accordance to NIOSH 42 CFR Part 84; EN 12941; CE 1094 (EVA PAPR with Loose Fitting Headtops only) requirements

3. Bullard® PAPRFC5 AM/FM/MA/AG/HE Filter for EVA PAPR:

Bullard HEPA Filter is designed to work with the EVA series PAPR system

Key Features:
• Filter Type: Combination Mechanical & Chemical
• Protection: Particulates, Ammonia, Formaldehyde, Methylamine, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, and Hydrogen Fluoride
• Particulate Efficiency: High Efficiency Particulate Absolute (HEPA) 99.99% vs. a mass mean 0.3 micron aerosol at 85 LPM Media secured with urethane glue around perimeter
• Filter Housing Material: ABS Plastic
• Sorbent Bed: Activated Carbon (Chromium Free)

Certifications:
NIOSH 42 CFR Part 84 TC-23C-2886 (EVA PAPR with Loose Fitting Headtops)
56,100.00 - ₹183,000.00INR / Pkg of 6

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Bullard PAPRFC3  Filter Cartridge for EVA-Particulate Protection; 6/Pk 1635920 Mfr # PAPRFC3
COMPARE
Bullard PAPRFC3 Filter Cartridge for EVA-Particulate Protection; 6/Pk
₹56,100.00INR / Pkg of 6
Bullard PAPRFC4  Filter Cartridge for EVA-Particulate and Organic Vapour Protection; 6/Pk 1635921 Mfr # PAPRFC4
COMPARE
Bullard® PAPRFC4 Filter Cartridge for EVA-Particulate and Organic Vapour Protection; 6/Pk
₹131,000.00INR / Pkg of 6
Bullard PAPRFC5 Filter Cartridge for EVA-Particulate and Hazardous Chemicals Protection; 6/Pk 1635922 Mfr # PAPRFC5
COMPARE
Bullard® PAPRFC5 Filter Cartridge for EVA-Particulate and Hazardous Chemicals Protection; 6/Pk
₹183,000.00INR / Pkg of 6
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat