Buck Scientific IR Spectroscopy Accessories

Optimize performance of your IR spectrophotometer
  • Choose from a wide range of parts and accessories compatible with your IR spectrophotometer
Expand the testing capabilities of your IR spectrometer and make sure it's running at its best.
11,600.00 - ₹82,500.00INR / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Buck Scientific 6401 Beta Cell for Infrared (IR) Spectroscopy; 25 mm Windows Not Included 8300100 Mfr # 6401
COMPARE
Beta Cell for Infrared (IR) Spectroscopy; 25 mm Windows Not Included
₹26,400.00INR / Each
Buck Scientific 6109 Nujol Mineral Oil for IR Spectroscopy; 2 oz 83001-15 8300115 Mfr # 6109
COMPARE
Nujol Mineral Oil for IR Spectroscopy; 2 oz 83001-15
₹11,600.00INR / Each
Buck Scientific 5231 Potassium Bromide (KBr) Powder for IR Spectroscopy; 100 grams 8300125 Mfr # 5231
COMPARE
Potassium Bromide (KBr) Powder for IR Spectroscopy; 100 grams
₹61,400.00INR / Each
Buck Scientific 500A3 NaCl Salt Window for IR Spectroscopy; 25 mm Diameter x 4 mm Thick 8300156 Mfr # 500A3
COMPARE
NaCl Salt Window for IR Spectroscopy; 25 mm Diameter x 4 mm Thick
₹13,900.00INR / Each
Buck Scientific 6106 Bench Wrench Kit for IR Spectroscopy 8300252 Mfr # 6106
COMPARE
Bench Wrench Kit for IR Spectroscopy
₹18,500.00INR / Each
Buck Scientific 6801 Beta Gas Cells for Gas Samples; 10 cm pathlength 8300400 Mfr # 6801
COMPARE
Beta Gas Cells for Gas Samples; 10 cm pathlength
₹82,500.00INR / Each
Buck Scientific 6806 Beta Gas Cell Replacement Body, for 10 cm Pathlength Cells 8300450 Mfr # 6806
COMPARE
Beta Gas Cell Replacement Body, for 10 cm Pathlength Cells
₹27,900.00INR / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat