Buck Scientific 6401 Beta Cell for Infrared (IR) Spectroscopy; 25 mm Windows Not Included
Representative image only.
Buck Scientific 6401 Beta Cell for Infrared (IR) Spectroscopy; 25 mm Windows Not Included
Buck Scientific | Mfr: 6401| Item: 8300100
Optimize performance of your IR spectrophotometer
₹26,400.00INR / Each
₹26,400.00INR / Each
Buy It Together
Current Item
Buck Scientific 6401 Beta Cell for Infrared (IR) Spectroscopy; 25 mm Windows Not Included
Buck Scientific 6401 Beta Cell for Infrared (IR) Spectroscopy; 25 mm Windows Not Included
8300100
₹26,400.00INR / Each
+ =
Total Price 40300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Choose from a wide range of parts and accessories compatible with your IR spectrophotometer
More About this Item
Expand the testing capabilities of your IR spectrometer and make sure it's running at its best.
Ideal for smears, this economical cell is easy to assemble. Simply place sample between two plain windowa separated by spacers, place in holder, and install threaded retainer until assembly is sealed. Requires two NaCl windows (sold separately).
Includes removable cell mount, 12 assorted PTFE spacers, slide plate holder, and instructions.
Specifications
  • DescriptionBeta Cell for Infrared (IR) Spectroscopy; 25 mm Windows Not Included