Buck Scientific 6801 Beta Gas Cells for Gas Samples; 10 cm pathlength

Representative image only.

Buck Scientific 6801 Beta Gas Cells for Gas Samples; 10 cm pathlength

Buck Scientific | Mfr: 6801| Item: 8300400

Optimize performance of your IR spectrophotometer

₹82,500.00INR / Each

₹82,500.00INR / Each

Buy It Together

Current Item

Buck Scientific 6801 Beta Gas Cells for Gas Samples; 10 cm pathlength
Buck Scientific 6801 Beta Gas Cells for Gas Samples; 10 cm pathlength
8300400

₹82,500.00INR / Each
+ =
Total Price 96400.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Choose from a wide range of parts and accessories compatible with your IR spectrophotometer

More About this Item

Expand the testing capabilities of your IR spectrometer and make sure it's running at its best.

Disassemble and reassemble these leaktight gas cells quickly and easily. Use the 5 cm gas cell with contentrated gases and 10 cm cell for less concentrated gases. Both feature a PYREX(R) body with two stems fitted with replaceable rubber septa. Window seals are made with VITON(R) O-rings; order PTFE O-rings separately. Cells require two NaCl windows.

Specifications
  • DescriptionBeta Gas Cells for Gas Samples; 10 cm pathlength
Reviews
Q&A