Bourdon Tube Pressure Gauges, Stainless Steel
Meets American Petroleum Institute (API) specifications
  • Suitable for corrosive environments, and gaseous or liquid media
  • Excellent load-cycle stability and shock resistance
  • All stainless steel construction with safety glass window
These all stainless steel gauges are designed to meet API and industry-specific specifications. Rugged construction makes these pressure gauges well suited for tough process industries including chemical, petroleum/refining, power stations, onshore and offshore mining, environmental technology, and new plant construction.
7,200.00 - ₹16,200.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accuracy Liquid Fill Max Pressure (PSI)
Availability Pricing
Wika 9678000 Hydraulic Pressure Gauge, 0 to 100 psi, 2.5 Dial, 1/4" NPT(M) 6843413 Mfr # 9678000
COMPARE
±2.5% full-scale Glycerine 100
₹7,200.00INR / Each
Wika 9768343 Bourdon Tube Pressure Gauge, 0 to 30 psi, 2.5" Dial, 1/4 NPT(M) 6843438 Mfr # 9768343
COMPARE
±2.5% full-scale Prepared for filling 30
₹15,600.00INR / Each
Wika 9768718 Bourdon Tube Pressure Gauge, 0 to 60 psi, 2.5" Dial, 1/4 NPT(M) 6843482 Mfr # 9768718
COMPARE
±2.5% full-scale Prepared for filling 60
₹13,700.00INR / Each
Wika 9833557 Bourdon Tube Pressure Gauge, 0 to 200 psi, 2.5" Dial, 1/4 NPT(M) 6843483 Mfr # 9833557
COMPARE
±2.5% full-scale Glycreine 200
₹14,700.00INR / Each
Wika 9833493 Bourdon Tube Pressure Gauge, 0 to 2000 psi, 2.5" Dial, 1/4" NPT(M) 9807808 Mfr # 9833493
COMPARE
±2.5% full-scale Glycerine 2000
₹16,200.00INR / Each
Wika 9833646 Bourdon Tube Pressure Gauge, 30" Hg to 0 psi, 2.5" Dial, 1/4" NPT(M) 9807810 Mfr # 9833646
COMPARE
±2.5% full-scale Glycerine 0
₹15,400.00INR / Each