Azlon Cylinders, Squat Form, PP

Squat form base allows more stability and saves space on working area
Manufactured of translucent polypropylene, these unbreakable cylinders feature blue enameled graduations from easy reading. Anti suction base eliminates problems when working on wet surfaces. Cylinders also feature unique self-draining base that allows wastewater to drain when using a labortory washing machine; helps eliminate contamination and spill re-wetting. Autoclavable.
5,900.00 - ₹71,100.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Azlon 537835-4000 Squat-Form PP Graduated Cylinders, 4000 mL, 1/Pk WW-06137-08 Mfr # 537835-4000
COMPARE
4000 50 Polypropylene
₹71,100.00INR / Each
Azlon 537835-0100 Squat-Form PP Graduated Cylinders, 100 mL, 5/Cs WW-06137-30 Mfr # 537835-0100
COMPARE
100 2 Polypropylene
₹21,900.00INR / Case of 5
Azlon 537835-0250 Squat-Form PP Graduated Cylinders, 250 mL, 1/Pk WW-06137-32 Mfr # 537835-0250
COMPARE
250 5 Polypropylene
₹5,900.00INR / Each
Azlon 537835-0500 Squat-Form PP Graduated Cylinders, 500 mL, 1/Pk WW-06137-34 Mfr # 537835-0500
COMPARE
500 10 Polypropylene
₹7,600.00INR / Each
Azlon 537835-1000 Squat-Form PP Graduated Cylinders, 1000 mL, 1/Pk WW-06137-36 Mfr # 537835-1000
COMPARE
1000 20 Polypropylene
₹9,900.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat