AEMC Electrical Equipment Accessories

Choose from the following electrical accessories to ensure you electrical connections are safe and working properly.
11,700.00 - ₹151,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
AEMC 2130.60 Hand-Crank/Spooled Tape Measure; 100 Ft WW-20001-73 Mfr # 2130.60
COMPARE
Hand-Crank/Spooled Tape Measure; 100 Ft
₹11,700.00INR / Each
AEMC 2135.35 Test Kit for Ground Resistance Tester, 3 points, 150 ft WW-20015-09 Mfr # 2135.35
COMPARE
Test Kit for Ground Resistance Tester, 3 points, 150 ft
₹151,000.00INR / Each
AEMC 2130.59 Calibration Checker, 25 Ohm WW-20033-77 Mfr # 2130.59
COMPARE
Accessory Calibration Checker, 25 Ohm
₹22,400.00INR / Each
AEMC MN93-BK AC Current Probe, 240A, 10 ft Leads WW-20034-01 Mfr # 2140.32
COMPARE
AC Current Probe, 240A, 10 ft Leads
₹56,200.00INR / Each
AEMC 193-24-BK Ampflex sensor, 24 in., 6500 A, 10ft Leads WW-20034-02 Mfr # 2140.34
COMPARE
Ampflex sensor, 24 in., 6500 A, 10ft Leads
₹75,500.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat