AEMC 193-24-BK Ampflex sensor, 24 in., 6500 A, 10ft Leads

Representative image only.

AEMC 193-24-BK Ampflex sensor, 24 in., 6500 A, 10ft Leads

AEMC | Mfr: 2140.34| Item: 2003402

₹99,100.00INR / Each

₹99,100.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Choose from the following electrical accessories to ensure you electrical connections are safe and working properly.

1 Year WarrantyConformite Europeenne
Specifications
  • DescriptionAmpflex sensor, 24 in., 6500 A, 10ft Leads
  • Warranty1 Year
Reviews
Q&A