AEMC MN93-BK AC Current Probe, 240A, 10 ft Leads

Representative image only.

AEMC MN93-BK AC Current Probe, 240A, 10 ft Leads

AEMC | Mfr: 2140.32| Item: 2003401

₹76,900.00INR / Each

₹76,900.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Choose from the following electrical accessories to ensure you electrical connections are safe and working properly.

Conformite Europeenne1 Year Warranty
Specifications
  • DescriptionAC Current Probe, 240A, 10 ft Leads
  • Warranty1 Year
Reviews
Q&A